missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ Microbiology Media Additives: Peptone
Additive for preparing bacterial nutrient media
£17.84 - £118.63
Specifications
Form | Powder |
---|---|
Grade | Molecular genetics |
Media Type | Peptone |
Product Type | Peptone |
For Use With (Application) | Nitrogen in a form readily available to bacteria |
Product Code | Brand | Packaging Type | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Packaging Type | Quantity | Price | Quantity & Availability | ||||
10296044
|
Fisher BioReagents™
BP1420-2 |
Poly Bottle | 2000 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10286764
|
Fisher BioReagents™
BP1420-100 |
Poly Bottle | 100 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
11413603
|
Fisher BioReagents™
BP1420-500 |
Poly Bottle | 500 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Dehydrated.
Specifications
Powder | |
Peptone | |
Nitrogen in a form readily available to bacteria |
Molecular genetics | |
Peptone | |
Poly Bottle |