missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ Taq DNA Polymerase

Allows up to 100 cycles of amplification without loss of activity

Brand:  Fisher BioReagents™ FB6000-35

Code : 32

Additional Details : CAS Number : 9012-90-2 Weight : 0.00001kg

 View more versions of this product

Product Code. 10698393

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

  • Licensed for use in PCR
  • Uses a thermostable DNA polymerase that has been encoded by a modified form of Thermus aquaticus DNA polymerase gene that has been inserted into an E. coli host
  • Has a 5«→3« exonuclease activity
  • Available with two different reaction buffers
  • Buffer A contains an optimized final concentration of 15mM MgCl2 for most PCR amplifications
  • Buffer B does not contain MgCl2; MgCl2 is provided in a separate vial as a 25mM solution
  • Storage Buffer: 20mM Tris-HCl, pH 7.5 (at room temperature), 100mM KCl, 0.1mM EDTA, 1mM DTT, 50% Glycerol, 0.5% Tween™ 20, 0.5% Nonidet P40™
  • Recommended Assay Buffer: 10X Assay Buffer A (100mM Tris-HCl, pH 9.0 [at 25°C/77°F]; 500mM KCl; 15mM MgCl2) or 10X Assay Buffer B (100mM Tris-HCl, pH 9.0 [at 25°C/77°F]; 500mM KCl) and 25mM MgCl2 solution in a separate vial

Unit Definition: One unit of enzyme is defined as the amount that will incorporate 10nmol of dNTPs into acid-insoluble material per 30 minutes in a 10-minute incubation at 74°C (165°F) under the assay conditions above

Specifications

Specifications

10X assay buffer A : Tris-HCl 100mM, pH 9 at 25°C, KCl 500mM and MgCl2 15mM solution in a separate vial
Taq DNA Polymerase
Poly Tubes
Standard PCR
Tris-HCl 20mM, pH 7.5 (at room temperature), KCl 100mM, EDTA 0.1mM, DTT 1mM, Glycerol 50%, Tween™ 20 0.5%, Nonidet™ P40™ 0.5%
-20°C
Liquid
2500 Units
5U/μL
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Special Offers

Special Offers