Learn More
Fisher BioReagents™ exACTGene™ DNA Ladders
Ready-to-use DNA size standards are stable at room temperature—no need to thaw and refreeze
Brand: Fisher BioReagents™ BP2578-100
Code : 32
Additional Details : Weight : 0.01000kg
Description
Ready-to-use DNA size standards are stable at room temperature—no need to thaw and refreeze
- Specially engineered to provide blunt-end DNA of precise length for better band quality than classic markers
- Ideal for size assessment and quantitative determination of DNA mass
- Contain loading dye to reduce pipetting steps and save time
- Have two or three high intensity reference bands for better sizing
- Provided in sufficient quantity to load 100 lanes
Specifications
Colorless | |
Liquid | |
For general purpose, large digested DNA | |
100 Lanes | |
100 Lanes |
DNA Molecular Weight Marker/Ladder | |
Premixed with loading dye | |
13 | |
Stable at room temperature for two years. |