Learn More
Fisher BioReagents™ exACTGene™ Core Reagent Set B
Optimized for routine PCR applications
Brand:  Fisher BioReagents™ FB6245
Code : 32
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 10774724
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
Description
Description
  • Provide an economical alternative for high quality PCR
  • Kit components are also available separately
Specifications
Specifications
Fisher BioReagents™ exACTGene™ Complete PCR Kit
Core Reagent Set B
-20°C
Taq DNA Polymerase, 10X Buffer B, MgCl2, dNTPs
80U
Videos
Documentation