missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ EZ-Run™ Protein Gel Staining Solution

Ready-to-use solution for highly sensitive staining of proteins separated in polyacrylamide gels

£141.61 - £375.34

Specifications

Color Dark Blue
Product Type Stain
Form Liquid
Formulation Coomassie™ Brilliant Blue G-250
For Use With (Application) Highly sensitive staining of proteins separated in Polyacrylamide gels
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10786444
SDS View Documents Promotion Details
Fisher BioReagents™
BP3620-1
1 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10609933
SDS View Documents Promotion Details
Fisher BioReagents™
BP3620-4
4 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

EZ-Run Protein Gel Staining Solution contains Coomassie™ Brilliant Blue G-250. Due to the property of the dye to form colloidal particles, proteins are preferentially stained without significant background staining of the gel matrix.

  • Detection is linear over two orders of magnitude with as little as 5ng protein detected
  • Sensitivity: 5ng BSA in 10% SDS-PAGE, 1.0mm thick gel, well width 3mm
  • Does not contain toxic methanol or acetic acid
  • Provides rapid destaining with simple water washes
  • Can be reused twice without significant loss of sensitivity
  • Tested for staining sensitivity in minigels using purified proteins and Protein Molecular Weight Standards

Specifications

Specifications

Dark Blue
Liquid
Highly sensitive staining of proteins separated in Polyacrylamide gels
Stain
Coomassie™ Brilliant Blue G-250
Videos
SDS
Documents

Documents

Product Certifications