missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ EZ-Run™ Protein Gel Solutions

Resolution equal to or better than gradient precast gels

Brand:  Fisher BioReagents™ BP7712-500

Code : 32

Additional Details : CAS Number : 79-06-1 Weight : 0.55000kg Transport : UN number : 3426 Chem class : 6.1 Pack group : III


 View more versions of this product

Product Code. 10678393

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

A ready-to-use premixed solution of acrylamide, bisacrylamide, buffer and SDS that enables ultra-fine resolution of protein bands by denaturing PAGE. This novel formulation provides the resolution of a gradient gel with less preparative work than a standard discontinuous gel system.

  • Ready-to-pour EZ-Run Protein Gel Solution for 1-D or 2-D PAGE—just add APS and TEMED
  • Wide separation range—from 3 to 200kDa on same minigel
  • Continuous gel system—no stacking gel necessary
  • Proprietary (patent-pending) gel chemistry

EZ-Run Protein Gel Solution comes with an optimized proprietary running buffer; other traditional SDS-PAGE buffers should not be used with the EZ-Run gel solution.

Applications:

Compatible with post-electrophoresis applications, such as Western blotting, MALDI analysis, protein sequencing. Suitable for all commonly used gel staining methods including Coomassie™ blue, silver staining and fluorescent dyes.

Specifications

Specifications

Western blotting, MALDI analysis, Protein sequencing
12.5% protein gel solution
500mL
3 to 200 kd
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 12.5% EZ-Run Protein Gel Solution with buffer

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 1B
  • Reproductive toxicity Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H372-Causes damage to organs.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H361f-Suspected of damaging fertility.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

SDS Documentation