missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ EZ-Run™ Rec Protein Ladder

Mixture of 14 highly purified recombinant proteins optimized for precise sizing of proteins by SDS-PAGE and Western blotting

Brand:  Fisher BioReagents™ BP3602-1

Code : 32

Additional Details : Weight : 0.32000kg

 View more versions of this product

Product Code. 10336693

  • £180.56 / Pack of 2


Estimated Shipment
16-03-2018
Add to basket

Specifications

Specifications

Molecular weight determination of polypeptides analyzed in various Gel system
<51% Glycerin, 2% Sodium Lauryl Sulfate, <1% Tris 9Hydroxymethyl) Aminomethane, <0.76% 2,3-Butanediol, 1,4-Dimercapto, <0.5% Sodium Chloride, <0.05% Ethylenediaminet
−20°C
10 to 200 kd
Supplied in gel-loading buffer
2 × 500μL
200 Loadings
Videos
SDS Documentation