Fisher BioReagents™ Bovine Serum Albumin, Fraction V, Heat Shock Treated

Suitable for immunological studies

Overview
Brand: Fisher BioReagents™

Manufacturer Part Number: BP1600-100

UNSPSC: 12140000

Code: NEW

Additional Details:
Additional Details: CAS Number: 9048-46-8 Weight: 0.10000kg



Product Code. 11493823

Quantity Price
1 £ 179.2 / 100g
EU Stock 1
Add to basket


Description and Specification

Specification

For Use With (Application) Immunological Studies
Quantity 100g
Source Bovine
Name BSA, Fraction V, Heat Shock Treated
Packaging Poly Bottle
pH Range 7 ±0.3