missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Safety Products™ Fireman Glove

Catalog Number p-7115907
Click to view available options
Size:
8
11
This item is not returnable. View return policy
This item is not returnable. View return policy

Water repellent cowhide grain leather, split leather, and Kevlar™ are combined to make this thermally and mechanically resistant glove. Honeywell Safety Products™ Fireman Glove is designed with an elasticized back as well as a velcro strap for a secure fit.

  • Heat resistant glove (formerly used as glove for firemen).
  • Yellow water-repellent cowhide grain leather glove.
  • Gunn cut, winged thumb, gauntlet type.
  • Hand lined with Kevlar™ cotton jersey for comfort and protection from cuts and thermal hazards.
  • Stitched with Kevlar™ thread for thermal insulation and sturdy seam resistance to heat and wear.
  • Elasticated back.
  • Velcro strap.
  • Split leather cuff.
TRUSTED_SUSTAINABILITY
Certifications/Compliance CE CATEGORY III Irreversible risks, EN 420-2003 EEC Basic standards, EN 388-2003 Mechanical hazards, EN 407-2004 Thermal hazards, EC Category PPE 2, EC Certificate Number 0075/797/162/12/09/0517
Classification Dexterity: 5
Cut Style Gunn
Cut Resistance Rating 2
Material Grain Leather, Cowhide Leather, Aramid, Cotton, Dupont™ Kevlar™ Aramid Fibers
Product Type Gloves
Size 8
Style Gauntlet
Thickness (Metric) 1.1 to 1.3 mm
For Use With (Application) Army-Defense and Fire Protection industries
Includes Velcro strap
Liner Material Dupont™ Kevlar™ aramid fiber, Cotton
Lining Lined
Puncture Rating 2
Quantity 10Pair/PK
Tear Rating 2
Thumb Style Wing
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'