Learn More
Duran™ FioLaX™ Test Tubes
Manufactured from borosilicate glass. DURAN™ FioLaX™ features a beaded rim.
Brand:  Duran™ 261102804
Code : A0
Additional Details : Weight : 1.40000kg
 View more versions of this product
Product Code. 10662781
  • £51.00 / Pack of 100
In Stock 38
Add to basket
Specifications
Specifications
Yes
Suited to Rapid Temperature Changes or Localized Heating
Yes
Borosilicate Glass
20mm
100Pack
45mL
Test Tube
Videos