Learn More
Thermo Scientific™ Finnpipette™ F1 Variable Volume Pipettes
The Finnpipette™ F1 system delivers performance with precision, all in a lightweight design that maximizes user comfort and reduces strain. It’s the ideal system for applications using expensive reagents and valuable samples.
Brand:  Thermo Scientific™ 4641040N
Additional Details : Weight : 0.27000kg
 View more versions of this product
Product Code. 15561885
  • £289.70 / Each
Estimated Shipment
05-11-2020
Add to basket
Description
Description
For decades, Thermo Scientific Finnpipette pipetting systems have delivered unmatched productivity and ergonomics across thousands of labs and applications worldwide.
Finnpipette F1: Everything you want in your everyday, go-to pipette
Ergonomically In-touch
Lightweight handle design combined with low plunger, tip attachment and ejection forces help reduce the risk of RSI
Silent, Effortless Volume Adjustment
Enjoy the new silent and lighter volume adjustment mechanism with over 50% lower forces compared to earlier models.
Secure Volume Lock
The secure volume setting prevents undesirable volume drift during operation. Simply lift the plunger up to set volume and press down to lock in place with minimum effort.
Adjustable Comfort
The 120° adjustable finger rest – for both right- and left-handed operator comfort – allows the hand to relax during pipetting cycles.
Super Blow-out
150% increase in air boost to ensure efficient delivery of micro-volumes and prevent capillary action in 50μl models and below.
Wide Selection of Finntip Pipette Tips
Finntips are designed and manufactured expressly for Finnpipette models, enabling optimal performance, precision and accuracy.
CE marked
The Finnpipette F1 Pipette together with compatible Finntip tips is CE marked in accordance with IVD Directive in Europe.
5 Year Warranty
With convenient online registration, and may vary by country.
Learn more about the complete Finnpipette family >
Specifications
Specifications
Finntip 250, Finntip Flex 200
0.02μL
1 to 10 μL
Finnpipette F1
Yellow