missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Finnpipette™ F1 Good Laboratory Pipetting (GLP) Kits

For your convenience, we packaged a variety of Finnpipette™ F1 single channels in a range of volumes, compatible Finntip pipette tips and a pipette stand to support good laboratory pipetting in your lab

Brand:  Thermo Scientific™ 4701060N

Additional Details : Weight : 3.03000kg

 View more versions of this product

Product Code. 15571945

  • £672.22 / Each


Estimated Shipment
09-12-2020
Add to basket

Description

Description

For decades, Thermo Scientific Finnpipette pipetting systems have delivered unmatched productivity and ergonomics across thousands of labs and applications worldwide.

Finnpipette F1: Everything you want in your everyday, go-to pipette

Ergonomically In-touch
Lightweight handle design combined with low plunger, tip attachment and ejection forces help reduce the risk of RSI

Silent, Effortless Volume Adjustment
Enjoy the new silent and lighter volume adjustment mechanism with over 50% lower forces compared to earlier models.

Secure Volume Lock
The secure volume setting prevents undesirable volume drift during operation. Simply lift the plunger up to set volume and press down to lock in place with minimum effort.

Adjustable Comfort
The 120° adjustable finger rest – for both right- and left-handed operator comfort – allows the hand to relax during pipetting cycles.

Super Blow-out
150% increase in air boost to ensure efficient delivery of micro-volumes and prevent capillary action in 50μl models and below.

Wide Selection of Finntip Pipette Tips
Finntips are designed and manufactured expressly for Finnpipette models, enabling optimal performance, precision and accuracy.

CE marked
The Finnpipette F1 Pipette together with compatible Finntip tips is CE marked in accordance with IVD Directive in Europe.

5 Year Warranty
With convenient online registration, and may vary by country.

Learn more about the complete Finnpipette family >
Specifications

Specifications

Finntip Flex tips: 200; 1000μL
Finnpipette F1 GLP Kit 4: 2-20μL, 20-200μL and 100-1000μL
2 to 1000 μL
Videos
SDS
Documents