Learn More
Thermo Scientific™ Filters and Accessories for Thermo Scientific Multiskan™ FC Microplate Readers
Multiskan™ FC microplate photometers can be equipped with wide range of additional filters to expand the instrument functionality and application flexibility.
Brand: Thermo Scientific™ 1426305
Code : A0
Additional Details : Weight : 0.02000kg
Description
Multiskan FC brings together over 40 years' experience of the well-known Multiskan product brand, with features for enhanced usability. The Multiskan FC has a 340–850 nm wavelength range, enabling a wide variety of applications from enzyme kinetic studies to Lowry assays. Extensive choice of filters for optimal wavelength selection for assays increases the application flexibility.
- Multiskan FC is equipped with an eight-position filter wheel with three preinstalled standard filters, 405 nm, 450 nm and 620 m.
- A comprehensive range of easy-to install additional filters is available for order
- Superior usability and logical workflow with Thermo Scientific™ SkanIt™ Software
Specifications
Optical Filter | |
Multiskan FC | |
Multiskan™ | |
630 nm |