Learn More
Integra Biosciences™ Pipetboy Filter
Prevent contamination with replaceable sterile filters. Integra Biosciences™ Pipetboy Filters are designed for use with Pipetboy Pro pipette controllers.
Brand:  Integra Biosciences™ 156607
Code : 04
Additional Details : Weight : 0.00094kg
 View more versions of this product
Product Code. 11656580
  • £5.50 / Each
Estimated Shipment
07-03-2018
Add to basket
Description
Description
  • Spare filter
  • Accessory for Pipetboy Pro Pipette controllers
Specifications
Specifications
0.2μm
No
Filter
Videos
Documentation
Technical Documents