Learn More
GVS Filter Technology SPR404 Elipse Integra Safety Goggle
his mask is recommended for dust and fumes containing substances. Additionally, the filters remove small amounts of VOC characterised by smell/nuisance odor.
Brand: GVS SPR404
This item is not returnable.
View return policy
Description
This mask is recommended for dust and fumes containing substances such as micro-organisms, marble, gypsum, titanium oxide, soapstone, rock wool, wood, detergents, textile fibers, spices, salt, animal feeds, etc..
- Protects against dust that can cause lung disease. In particular, protects against coal, silica, cotton, iron ore, graphite, kaolin, zinc, aluminium dusts.
- Protects against harmful dusts such as asbestos, bauxite, coal, silica, iron, and against toxic dusts such as manganese, lead and chromium.
- Additionally the filters remove small amounts of VOC characterised by smell/nuisance odor.
- Extremely lightweight with integrated eye and respirator protection to give a better face fit, more comfort, and reduce the risk of mist building up inside the lens.
- Low breathing resistance to reduce the risk of user fatigue and to add additional comfort.
- Made from soft thermoplastic odorless elastomer that is hypoallergenic (without latex and silicone) and an easy-to-adjust headband.
Specifications
Blue | |
Reusable |