missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Filter for Recirculating Air Storage Cabinet
Activated carbon filter for air filtration storage cabinets
£391.14 - £429.30
Specifications
Depth (Metric) Exterior | 270 mm |
---|---|
Product Type | Activated Carbon Filter |
Includes | FX-CX |
Width (Metric) Exterior | 270 mm |
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
15987556
|
asecos™
32950 |
Model FX90 |
£429.30
Each |
Estimated Shipment: 15-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15997556
|
asecos™
32951 |
Model FX90 & FX30 |
£391.14
Each |
Estimated Shipment: 15-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Filter
- Activated carbon
- For models: FX90 and/or FX30
- For tall cabinets
- Dimensions exterior (W x D): 270 x 270 mm
Specifications
270 mm | |
Activated Carbon Filter | |
FX-CX | |
270 mm |