Learn More
Fisherbrand™ Filter Pump with Non-Return Valve
Fitted with rifled nozzles for water and vacuum connections.
Brand: Fisherbrand™
Code : NEW
Additional Details : Weight : 0.50000kg
Description
- Manufactured in nickel-plated brass and stainless steel
- Exceptionally robust design
Specifications
0.5 in. BSP connecting thread | |
Water and vacuum connections | |
Filter pump | |
Non-return valve |