missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VICI Jour Chromatography™ Filter in-line PEEK
Brand: VICI Jour Chromatography™ 724JR68256
Code : 15
Additional Details : Weight : 1.00000kg
Specifications
Encased Titanium Frit 5μm | |
5 μm | |
30 mL/min | |
12.7 μL |