missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Robu™ Filter Funnel
Robu™ Filter Funnels are designed to be use with a Buchner funnel.
Brand: Robu™ 21120
Additional Details : Weight : 0.10000kg
Description
- Can withstand acids, alkalis, and solvents and even temperatures of up to 540°C
- Filter sucking
- Form according to Büchner
Specifications
Filter Funnel | |
125 mL | |
Borosilicate Glass | |
160 to 250 μm |
65 mm outer, 60 mm inner | |
10 mm | |
56 mm |