Learn More
Ebro Electronic™ AH 300 Sinter-Filter (stainless steel)
The stainless steel sintered filter AH 300 also increases the protection class to IP 40.
Brand: Ebro Electronic™ 1340-5625
Code : A0
Additional Details : Weight : 0.80000kg
Description
The stainless steel massively increases the mechanical protection, e.g. from crushing the filter. Nothing will happen to your probe! In addition, the stainless steel is highly resistant to corrosion even in high temperature applications.Specifications
Filter | |
Stainless Steel | |
TFH610/TFH620 Hygrothermometer | |
1 Each |