missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Filaggrin Mouse, Clone: FLG/1561, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 2312-MSM1-P1
Additional Details : Weight : 0.01000kg
Specifications
Filaggrin | |
Cytoplasmic | |
FLG/1561 | |
Unconjugated | |
NP_002007 | |
Mouse | |
26-45kDa (Processed) ;350kDa (Profilaggrin) | |
Protein A/G Purified | |
RUO | |
2312 | |
IgG1 |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
FLG | |
Recombinant human Filaggrin protein fragment (aa 998-1104) | |
U-251-MG cells or Skin | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |