missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Field's Stain B
Click to view available options
Quantity:
10g
50g
Description
Used to stain blood smears to identify malarial parasites
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Molecular Formula | C20H6Br4Na2O5 |
InChI Key | SEACYXSIPDVVMV-UHFFFAOYSA-L |
Product Type | Stain |
SMILES | [Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=C(Br)C(=O)C(Br)=C2OC2=C(Br)C([O-])=C(Br)C=C12 |
IUPAC Name | disodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate |
Quantity | 10g |
Molecular Weight (g/mol) | 691.86 |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'