missing translation for 'onlineSavingsMsg'
Learn More

Ficoll 400 (White to Off-White Powder/Molecular Biology), Fisher BioReagents

Brand:  Fisher Bioreagents BP525-5

Code : 32

Additional Details : CAS Number : 26873-85-8 Weight : 0.05500kg

Packaging Poly Bottle
Quantity 5g

 View more versions of this product

Product Code. 10562945

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Description

Description

Ficoll 400 is used as a component of Denhardt's Reagent and in gel-loading buffers for DNA gels. It is also used in forming density gradients to separate cells and subcellular components.
Specifications

Chemical Identifiers

Ficoll™ 400
Molecular Biology Grade
≈400,000
26873-85-8
Powder/Solid

Specifications

Ficoll™ 400
Molecular Biology Grade
Powder/Solid
Poly Bottle
Inclusive between 50° (+ or -)
5g
26873-85-8
DNase free
≈400,000
Protease free
Off-white
Videos
SDS Documentation