missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ficoll|r 400, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
Ficoll™ 400 is used for cell separation and organelle isolation. It is also used to separate blood to its components like erythrocytes and leukocytes. It acts as a mononuclear cell separator and used in culture media, nucleic acidhybridization, electrophoresis and in immunological studies.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Solubility Information | Soluble in water. |
Quantity | 10 g |
Formula Weight | ca 400,000 |
Chemical Name or Material | Ficoll 400 |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'