missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ficolin-2 Mouse anti-Human, Clone: 297018, R&D Systems™
Mouse Monoclonal Antibody
POA
Specifications
Antigen | Ficolin-2 |
---|---|
Clone | 297018 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human Ficolin-2 Leu26-Ala313 Accession # Q15485 |
Gene ID (Entrez) | 2220 |
Gene Accession No. | Q15485 |
Description
Ficolin-2 Monoclonal specifically detects Ficolin-2 in Human samples. It is validated for Western Blot.Specifications
Ficolin-2 | |
Mouse myeloma cell line NS0-derived recombinant human Ficolin-2 Leu26-Ala313 Accession # Q15485 | |
Q15485 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Mouse | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
RUO |
297018 | |
2220 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Primary | |
Detects human Ficolin-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Ficolin-3 is observed. | |
Unconjugated | |
Western Blot | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only