missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Fibrinogen-Binding Peptide

For cellular and molecular biology applications

Brand:  Alfa Aesar™ J61540.MB

Code : NEW

Additional Details : CAS Number : 137235-80-4 Weight : 0.00025kg

 View more versions of this product

Product Code. 15444129

  • £204.79 / 25mg


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Disclaimers:

For Research Use Only


Specifications

Specifications

565.63
25mg
Powder
RUO
Fibrinogen-Binding Peptide
HPLC
Antithrombotic agent
H-Glu-His-Ile-Pro-Ala-OH
2° to 8°C
Videos
SDS