missing translation for 'onlineSavingsMsg'
Learn More

FH 535, Tocris Bioscience™

Inhibitor of Wnt/β-catenin signaling

Brand:  RD Systems (Bio-Techne) 4344/50

Code : Z1

Additional Details : Weight : 0.00050kg

Quantity 50mg

 View more versions of this product

Product Code. 13444594

  • £324.73 / 50mg

Estimated Shipment
08-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Inhibitor of Wnt/β-catenin signaling and dual antagonist of PPARγ/δ activity. Suppresses β-catenin/Tcf-mediated transcription; inhibits β-catenin and GRIP1 recruitment to PPARγ and δ. Exhibits antiproliferative effects in transformed colon, lung and liver cancer cell lines.
Specifications

Chemical Identifiers

108409-83-2
361.193
2,5-dichloro-n-2-methyl-4-nitrophenyl benzenesulfonamide, beta-catenin/tcf inhibitor, fh535, n-2-methyl-4-nitrophenyl-2,5-dichlorobenzenesulfonamide, cbmicro_011295, fh hplc
2,5-dichloro-N-(2-methyl-4-nitrophenyl)benzenesulfonamide
C13H10Cl2N2O4S
AXNUEXXEQGQWPA-UHFFFAOYSA-N
3463933
CC1=C(C=CC(=C1)[N+](=O)[O-])NS(=O)(=O)C2=C(C=CC(=C2)Cl)Cl

Specifications

FH 535
108409-83-2
2,5-dichloro-n-2-methyl-4-nitrophenyl benzenesulfonamide, beta-catenin/tcf inhibitor, fh535, n-2-methyl-4-nitrophenyl-2,5-dichlorobenzenesulfonamide, cbmicro_011295, fh hplc
CC1=C(C=CC(=C1)[N+](=O)[O-])NS(=O)(=O)C2=C(C=CC(=C2)Cl)Cl
361.193
361.2
50mg
C13H10Cl2N2O4S
AXNUEXXEQGQWPA-UHFFFAOYSA-N
2,5-dichloro-N-(2-methyl-4-nitrophenyl)benzenesulfonamide
3463933
>99%
Videos