missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FGR Recombinant Human Protein
FGR is a SRC-related kinase present in hematopoietic cells.
£1602.00 - £6900.00
Specifications
Accession Number | NP_005239 |
---|---|
For Use With (Application) | Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology |
Formulation | 20mM Tris, pH 7.5, 100mM NaCl, 0.05mM EDTA, 0.05% NP-40, 1mM DTT, 50% glycerol |
Molecular Weight (g/mol) | 62kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15440303
|
Invitrogen™
PR4146B |
100μg |
£1602.00
100µg |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10158672
|
Invitrogen™
PV4078 |
1mg |
£6900.00
1mg |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
FGR signals within multiple pathways including within the integrin pathway and downstream of chemokine and cytokine receptors.
Specifications
NP_005239 | |
20mM Tris, pH 7.5, 100mM NaCl, 0.05mM EDTA, 0.05% NP-40, 1mM DTT, 50% glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
SRC Kinase Family | |
Recombinant | |
Baculovirus |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
62kDa | |
RUO - research use only | |
FGR | |
Human | |
His Tag | |
Frozen |