Learn More
FGIN-1-43, Tocris Bioscience™
Potent, specific ligand for mitochondrial DBI receptor
Brand:  Tocris (Bio-Techne) 065950
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13487853
  • £461.52 / 50mg
Estimated Shipment
03-10-2018
Add to basket
Description
Description
Most active of the 2-aryl-3-indoleacetamides; a new class of potent and specific ligands as probes for the mitochondial DBI receptor (peripheral benzodiazepine receptor).
Specifications
Chemical Identifiers
145040-29-5
487.509
N, N-Dihexyl-2-(4-chlorophenyl)-5-chloroindole-3-acetamide
C28H36Cl2N2O
XTZUPNNVXIMWAR-UHFFFAOYSA-N
3995234
Specifications
FGIN-1-43
145040-29-5
C28H36Cl2N2O
XTZUPNNVXIMWAR-UHFFFAOYSA-N
2-[5-chloro-2-(4-chlorophenyl)-1H-indol-3-yl]-N,N-dihexylacetamide
3995234
50mg
>98%
N, N-Dihexyl-2-(4-chlorophenyl)-5-chloroindole-3-acetamide
CCCCCCN(CCCCCC)C(=O)CC1=C(NC2=C1C=C(C=C2)Cl)C3=CC=C(C=C3)Cl
487.509
487.51
Videos