missing translation for 'onlineSavingsMsg'
Learn More

FGIN-1-27, Tocris Bioscience™

Potent, specific ligand for mitochondrial DBI receptor

Brand:  Tocris (Bio-Techne) 065810

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13457853

  • £99.96 / 10mg

Estimated Shipment date
from Supplier 31-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Binds specifically and with high affinity to the peptide DBI receptor (benzodiazepine receptor) on mitochondrial membranes, inducing production of neurosteroids, which modulate GABA receptors (EC50 = 3 nM for increase in production of pregnenolone in glial cells). Thus it reduces anxiety without causing sedation.
Specifications

Chemical Identifiers

142720-24-9
436.615
fgin-1-27, fgin 1 27, n,n-dihexyl-2-4-fluorophenyl indole-3-acetamide, fgin 1-27, n,n-di-n-hexyl-2-4-fluorophenyl indole-3-acetamide, chembl63154, 1h-indole-3-acetamide, 2-4-fluorophenyl-n,n-dihexyl, 2-2-4-fluorophenyl-1h-indol-3-yl-n,n-dihexylacetamide, tocris-0658
2-[2-(4-fluorophenyl)-1H-indol-3-yl]-N,N-dihexylacetamide
C28H37FN2O
VUWXAQFLTSBUDB-UHFFFAOYSA-N
132496
CCCCCCN(CCCCCC)C(=O)CC1=C(NC2=CC=CC=C21)C3=CC=C(C=C3)F

Specifications

10mg
142720-24-9
fgin-1-27, fgin 1 27, n,n-dihexyl-2-4-fluorophenyl indole-3-acetamide, fgin 1-27, n,n-di-n-hexyl-2-4-fluorophenyl indole-3-acetamide, chembl63154, 1h-indole-3-acetamide, 2-4-fluorophenyl-n,n-dihexyl, 2-2-4-fluorophenyl-1h-indol-3-yl-n,n-dihexylacetamide, tocris-0658
CCCCCCN(CCCCCC)C(=O)CC1=C(NC2=CC=CC=C21)C3=CC=C(C=C3)F
436.615
436.61
FGIN-1-27
C28H37FN2O
VUWXAQFLTSBUDB-UHFFFAOYSA-N
2-[2-(4-fluorophenyl)-1H-indol-3-yl]-N,N-dihexylacetamide
132496
>98%
Videos
Documents