missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGFR2 Recombinant Human Protein
FGFR2 Recombinant Human Protein
Brand: Invitrogen PV3368
Code : 50
Additional Details : Weight : 0.05250kg
Description
FGFR2 is a member of the FGFR family of receptor tyrosine kinases. This family is known to regulate a host of cellular functions including angiogenesis, mitogenesis, osteogenesis, myogenesis, carcinogenesis, cellular differentiation, and tissue repair after injury. The FGFR family has also been implicated in a number of diseases including cancer, rheumatoid arthritis, and diabetic retinopathy.Specifications
NP_075261 | |
Pharma and Biopharma, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay | |
52.1kDa | |
10μg | |
For research use only. Not for use in diagnostic procedures | |
FGFR2, CEK3, TY14, BEK | |
FGFR2 was pre-diluted in enzyme dilution buffer and assayed in 12.5mM Tris (pH 7.5), 10mM MgCl2, 1mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2mM DTT, 0.01% Triton™ X-100, 200μM ATP, 200μg/mL poly [Glu, Tyr] 4:1 substrate and trace [32P]-γ-ATP for 10 minutes at 30°C | |
Amino acids 403-822 | |
2,700nmoles of phosphate transferred to poly [Glu, Tyr] 4:1 substrate per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 1μg/mL | |
His Tag | |
Kinase and inhibitors | |
85% by SDS-PAGE |
0.43mg/mL | |
2263 | |
FGFR2 Recombinant Human Protein | |
–80°C | |
1 Each | |
FGFR2 Recombinant Human Protein | |
FGF receptor | |
FGFR2 Recombinant Human Protein | |
Human | |
Baculovirus | |
Receptor tyrosine kinases |
For Research Use Only. Not for use in diagnostic procedures.