missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FGF-9 Antibody, R&D Systems™
Mouse Monoclonal Antibody
POA
Specifications
Antigen | FGF-9 |
---|---|
Clone | 36923 |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Classification | Monoclonal |
Description
FGF-9 Monoclonal specifically detects FGF-9 in Human samples. It is validated for Western Blot.Specifications
FGF-9 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
FGF9, FGF-9, Fibroblast growth factor 9, fibroblast growth factor 9 (glia-activating factor), GAF, glia-activating factor, HBFG-9, HBGF-9, Heparin-binding growth factor 9, MGC119914, MGC119915, SYNS3 | |
FGF9 | |
IgG1 | |
Protein A or G purified from ascites | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
36923 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P31371 | |
2254 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human FGF-9 Met1-Ser208 Accession # P31371 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human FGF-9 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) FGF-3, -4, -5, -7, -10, -11, -13, -16, -17, -18, -19, rhFGF acidic, rhFGF basic, recombinant mouse (rm) FGF-6, -8b, -8c, or -15 is observed. In Western blots, no cross-reactivity with rhFGF-3, -4, -5, -6, -7, -10, -13, rmFGF-8b, or rmFGF-8c is observed. |
For Research Use Only