missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse FGF-6 Antibody, R&D Systems™
Sheep Polyclonal Antibody
POA
Specifications
Antigen | FGF-6 |
---|---|
Concentration | LYOPH |
Dilution | Immunohistochemistry 5-15 ug/mL |
Applications | Immunohistochemistry |
Classification | Polyclonal |
Description
FGF-6 Polyclonal specifically detects FGF-6 in Mouse samples. It is validated for Immunohistochemistry.Specifications
FGF-6 | |
Immunohistochemistry 5-15 ug/mL | |
Polyclonal | |
Sheep | |
Mouse | |
FGF6, FGF-6, fibroblast growth factor 6, HBGF-6, Heparin secretory-transforming protein 2, Heparin-binding growth factor 6, HST2, HST-2, HSTF2, HSTF-2 | |
FGF6 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
NP_034334 | |
2251 | |
E. coli-derived recombinant mouse FGF-6 Pro39-Ile208 Accession # NP_034334 | |
Primary | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Detects mouse FGF-6 in direct ELISAs. In direct ELISAs, approximately 15% cross-reactivity with recombinant human FGF-6 and less than 1 % cross-reactivity with recombinant mouse FGF-7 is observed. |
For Research Use Only