missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse FGF-4 Antibody, R&D Systems™
Sheep Polyclonal Antibody
POA
Specifications
Antigen | FGF-4 |
---|---|
Concentration | LYOPH |
Dilution | Immunohistochemistry 5-15 ug/mL |
Applications | Immunohistochemistry |
Classification | Polyclonal |
Description
FGF-4 Polyclonal specifically detects FGF-4 in Mouse samples. It is validated for Immunohistochemistry.Specifications
FGF-4 | |
Immunohistochemistry 5-15 ug/mL | |
Polyclonal | |
Sheep | |
Mouse | |
FGF4, fibroblast growth factor 4, HBGF-4, HBGF-4Transforming protein KS3, heparin secretory transforming protein 1, Heparin secretory-transforming protein 1, Heparin-binding growth factor 4, HST-1, HST-1HSTF-1, HSTF1fibroblast growth factor 4 splice isoform, HSTFGF-4, human stomach cancer, transforming factor from FGF-related oncogene, kaposi sarcoma oncogene, KFGF, K-FGF, KS3, oncogene HST | |
FGF4 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
P11403 | |
2249 | |
E. coli-derived recombinant mouse FGF-4 Ala30-Leu202 Accession # P11403 | |
Primary | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Detects mouse FGF-4 in direct ELISAs. In direct ELISAs, approximately 60% cross-reactivity with recombinant human FGF-4 is observed. |
For Research Use Only