missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ FFN 200 dihydrochloride

Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
FFN 200 dihydrochloride is a selective fluorescent VMAT2 substrate. Selectively traces exocytosis in dopaminergic neuronal cell culture and brain tissue. Excitation and emission maxima are 352 and 451 nm, respectively.
Specifications
Specifications
| Chemical Name or Material | 4-(2-Aminoethyl)-7-amino-2H-1-benzopyran-2-one dihydrochloride |
| CAS | 2080306-27-8 |
| Quantity | 10 mg |
| Solubility | Soluble to 100mM in water and to 100mM in DMSO |
| Target | Fluorescent Transporter Probes |
| Molecular Formula | C11H12N2O20.2 HCl |
| Formula Weight | 277.15 |
| Physical Form | Solid |
| Color | White |
| Product Type | Bioactive Small Molecule |
| Show More |
For Research Use Only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction