Learn More
Gibco™ Fetal Bovine Serum, qualified, One Shot™ format
Gibco™ Fetal Bovine Serum, qualified, One Shot™ format offers excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.
Brand: Gibco™ A3160802
Additional Details : Weight : 1.00000kg
Description
- Wide mouth, chamfered edge, and curved shape for easy pouring and pipetting
- Engineered to freeze/thaw without splitting or breaking
- Fits in test tube racks and frees up freezer space
- No labeling mishaps with our easy-to-read package label containing all of the information you need
- 33% less waste generated than aliquoting from 500 ml bottles
Specifications
Store in freezer -5 to -30°C | |
South America | |
10 x 50 mL | |
Dry Ice |
Fewer resources used and less waste | |
One Shot | |
Qualified |