Ferrous Sulfate, 7-Hydrate, Crystal, Multi-Compendial, J.T.Baker™


Overview
Brand: JT Baker

Manufacturer Part Number: 2063-05

UNSPSC: 12000000

Additional Details:
Additional Details: Weight: 2.50000kg


Packaging Wide Mouth Amber Glass Bottle
Quantity 2.5kg

Product Code. 15517954

Quantity Price
1 £ 341.54 / 2.50kg
Call for latest availability
+44 (0)1509 555 500
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Ferrous Sulfate
CAS 7782-63-0
Name Note 7-Hydrate
Purity Grade Multi-Compendial
Molecular Formula FeSO4·7H2O
Formula Weight 278.02

Specification

Physical Form Crystals
Formula Weight 278.02
Purity Grade Multi-Compendial
Chemical Name or Material Ferrous Sulfate
CAS 7782-63-0
Molecular Formula FeSO4·7H2O
Name Note 7-Hydrate
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 1.90kg
Quantity 2.5kg