missing translation for 'onlineSavingsMsg'
Learn More

Ferrous Sulfate, 7-Hydrate, Crystal, Multi-Compendial, J.T.Baker™

Brand:  JT Baker 2063-01

Additional Details : CAS Number : 7782-63-0 Weight : 0.90000kg

Quantity 500g
Packaging Wide Mouth Amber Glass Bottle

 View more versions of this product

Product Code. 15507954

  • £109.00 / 500g


Estimated Shipment
15-05-2018
Add to basket

Specifications

Chemical Identifiers

Ferrous Sulfate
7-Hydrate
FeSO4·7H2O
7782-63-0
Multi-Compendial
278.02

Specifications

Ferrous Sulfate
7782-63-0
Crystals
278.02
1L = 1.90kg
7-Hydrate
Multi-Compendial
FeSO4·7H2O
Wide Mouth Amber Glass Bottle
500g
Videos