missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferrous Ammonium Sulfate, 6-Hydrate, Fine Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
419.90 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Wide Mouth Amber Glass Bottle
Specifications
Physical Form | Fine Crystals |
Quantity | 500 g |
Molecular Formula | Fe(NH4)2(SO4)2·6H2O |
Formula Weight | 392.13 |
Grade | BAKER ANALYZED™ ACS Reagent |
Packaging | Wide Mouth Amber Glass Bottle |
Chemical Name or Material | Ferrous Ammonium Sulfate, 6-Hydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'