Learn More
Ferroin Indicator Solution, Reagecon
High-quality indicator solution produced from the highest quality raw materials and manufactured and tested under rigorous conditions.
Brand: Reagecon PFS1
Code : NEW
Additional Details : Weight : 0.10000kg
Description
- Titrant, Standard Titrant, Standard Solution, Standardized Solution, Reference Standard, Reference Material
- For manual titrations; indicator titrant or analyte change color and this color change is by far the most important method of end point detection, in such titrations.
- A high quality indicator solution such as Reagecon's Ferroin Indicator Solution facilitates end point detection, in a clear and unambiguous manner and is an imperative for accurate manual titrimetry.
Specifications
Amber glass Winchester bottle | |
100 mL | |
Titration & Analyte Color Change | |
Liquid |