missing translation for 'onlineSavingsMsg'
Learn More

Ferrocenemethanol, 97%, ACROS Organics™

Brand:  Acros Organics 348030050

Code : 29

Additional Details : CAS Number : 1273-86-5 Weight : 0.05500kg

Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10410062

  • £352.70 / 5g


Estimated Shipment
26-02-2018
Add to basket

Specifications

Chemical Identifiers

Ferrocenemethanol
97%
C11H12FeO
MFCD00040878
1273-86-5
97%
215.93

Specifications

Ferrocenemethanol
77.0° to 82.0°C
1273-86-5
Glass bottle
97%
C11H12FeO
MFCD00040878
Yellow to gold to orange
5g
Authentic
Powder or Needles
97%
215.93
Videos
SDS