missing translation for 'onlineSavingsMsg'
Learn More

Ferrocenecarboxaldehyde, 98%, ACROS Organics™

Brand:  Acros Organics 153530250

UNSPSC : 12350000

Code : 29

Additional Details : CAS Number : 12093-10-6 Weight : 0.07500kg Transport : UN number : 3467 Chem class : 1 Pack group : II

Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10311424

  • £132.70 / 25g


EU Stock 1
Add to basket

Specifications Description & Specifications

Chemical Identifiers

Ferrocenecarboxaldehyde
≥97.5%
C11H10FeO
MFCD00001429
12093-10-6
98%
214.05
Cyclopentadienyl(formylcyclopentadienyl)iron

Specifications

Ferrocenecarboxaldehyde
115° to 122°C
12093-10-6
Glass bottle
≥97.5%
C11H10FeO
MFCD00001429
Orange to red to brown
25g
Authentic
Crystalline Powder or Crystals or Crystals
98%
214.05
Cyclopentadienyl(formylcyclopentadienyl)iron
Documentation

Technical Documents