Ferrocenecarboxaldehyde, 98%, ACROS Organics™


Overview
Brand: Acros Organics

Manufacturer Part Number: 153530050

UNSPSC: 12350000

Code: 29

Additional Details:
Additional Details: CAS Number: 12093-10-6 Weight: 0.06000kg Transport: UN number : 3467 Chem class : 1 Pack group : II


Packaging Glass bottle
Quantity 5g

Product Code. 10172960

Quantity Price
1 £ 38.3 / 5g
EU Stock 3
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Ferrocenecarboxaldehyde
CAS 12093-10-6
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C11H10FeO
Formula Weight 214.05
MDL Number MFCD00001429
Synonym Cyclopentadienyl(formylcyclopentadienyl)iron

Specification

Physical Form Crystalline Powder or Crystals or Crystals
Assay ≥97.5%
Formula Weight 214.05
Quantity 5g
Color Orange to red to brown
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
CAS 12093-10-6
MDL Number MFCD00001429
Assay Percent Range 98%
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
Molecular Formula C11H10FeO
Health Hazard 1 Danger
Synonym Cyclopentadienyl(formylcyclopentadienyl)iron
Chemical Name or Material Ferrocenecarboxaldehyde
Melting Point 115° to 122°C
Documentation

Technical documents

Safety and Handling

Danger

GHS H Statement:
Toxic if swallowed.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.