Ferrocenecarboxaldehyde, 98%, ACROS Organics™


Overview
Brand: Acros Organics

Manufacturer Part Number: 153530010

UNSPSC: 12350000

Code: 29

Additional Details:
Additional Details: CAS Number: 12093-10-6 Weight: 0.05100kg Transport: UN number : 3467 Chem class : 1 Pack group : II


Packaging Glass bottle
Quantity 1g

Product Code. 10147603

Quantity Price
1 £ 15.0 / 1g
EU Stock 3
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Ferrocenecarboxaldehyde
CAS 12093-10-6
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C11H10FeO
Formula Weight 214.05
MDL Number MFCD00001429
Synonym Cyclopentadienyl(formylcyclopentadienyl)iron

Specification

Assay ≥97.5%
Physical Form Crystalline Powder or Crystals or Crystals
Formula Weight 214.05
Quantity 1g
Color Orange to red to brown
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
CAS 12093-10-6
Assay Percent Range 98%
MDL Number MFCD00001429
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
Packaging Glass bottle
Health Hazard 1 Danger
Molecular Formula C11H10FeO
Chemical Name or Material Ferrocenecarboxaldehyde
Synonym Cyclopentadienyl(formylcyclopentadienyl)iron
Melting Point 115° to 122°C
Documentation

Technical documents

Safety and Handling

Danger

GHS H Statement:
Toxic if swallowed.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.