missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferric Oxide, Powder, BAKER ANALYZED™ Reagent, J.T. Baker™
Click to view available options
Quantity:
500 g
Packaging:
Wide Mouth Amber Glass Bottle
Specifications
Physical Form | Powder |
Quantity | 500 g |
Formula Weight | 159.69 |
Grade | BAKER ANALYZED™ Reagent |
Packaging | Wide Mouth Amber Glass Bottle |
Specific Gravity | 1L = 5.24kg |
Chemical Name or Material | Ferric Oxide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'