missing translation for 'onlineSavingsMsg'
Learn More

Ferric Nitrate, 9-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog Number p-5390753
260.30 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.


Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
This item is not returnable. View return policy
7782-61-8
FeH18N3O18
403.99
MFCD00149708
SZQUEWJRBJDHSM-UHFFFAOYSA-N
ferric nitrate nonahydrate, iron iii nitrate nonahydrate, unii-vmx4uop3vn, ferric nitrate, nonahydrate, iron iii nitrate, vmx4uop3vn, iron, reference standard solution, ferricnitratenonahydrate, nitric acid, iron 3+ salt, nonahydrate, iron iii nitrate hydrate
16211566
This item is not returnable. View return policy
7782-61-8
FeH18N3O18
403.99
MFCD00149708
SZQUEWJRBJDHSM-UHFFFAOYSA-N
ferric nitrate nonahydrate, iron iii nitrate nonahydrate, unii-vmx4uop3vn, ferric nitrate, nonahydrate, iron iii nitrate, vmx4uop3vn, iron, reference standard solution, ferricnitratenonahydrate, nitric acid, iron 3+ salt, nonahydrate, iron iii nitrate hydrate
16211566

High quality chemicals for laboratory and specialized industrial use

Physical Form Crystals
Quantity 500 g
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Fe+3]
IUPAC Name iron(3+);trinitrate;nonahydrate
Formula Weight 404
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Specific Gravity 1L = 1.68kg
Chemical Name or Material Ferric Nitrate, 9-Hydrate
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'