missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferric Nitrate, 9-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
260.30 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
Specifications
Physical Form | Crystals |
Quantity | 500 g |
SMILES | [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Fe+3] |
IUPAC Name | iron(3+);trinitrate;nonahydrate |
Formula Weight | 404 |
Grade | BAKER ANALYZED™ ACS Reagent |
Packaging | Poly Bottle |
Specific Gravity | 1L = 1.68kg |
Chemical Name or Material | Ferric Nitrate, 9-Hydrate |
Safety and Handling
Signal Word
- Danger
- Serious eye damage/eye irritation Category 2
- Oxidising solid Category 2
- Skin corrosion/irritation Category 2
- H272-May intensify fire; oxidiser.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'