missing translation for 'onlineSavingsMsg'
Learn More

Ferric Ammonium Sulphate R6 EU Pharmacopoeia, Fisher Chemical™

Catalog Number 12970414
Click to view available options
Quantity:
100 mL
Packaging:
Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.

Concentration 10μg/mL
CAS 74738-17-3
Packaging Bottle
Quantity 100 mL
Molecular Formula C11H6Cl2N2
InChI Key FKLFBQCQQYDUAM-UHFFFAOYSA-N
SMILES C1=CC(=C(C(=C1)Cl)Cl)C2=CNC=C2C#N
IUPAC Name 4-(2,3-dichlorophenyl)-1H-pyrrole-3-carbonitrile
Molecular Weight (g/mol) 237.083
PubChem CID 91724
ChEBI CHEBI:34759
Analyte or Component Names Ferric ammonium sulphate solution R6
Grade EU Ph Ref: 1037705
Show More Show Less
compliance-icons
Product Identifier
  • Ferric ammonium sulphate solution R6. European Pharmacopoeia Ref: 1037705 (RP 079)
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'