Fernbach Flask Clamps for Sartorius™ Certomat™ Benchtop Shakers

In stainless steel, for a variety of flask sizes

Overview
Brand: Sartorius Stedim Biotech

Manufacturer Part Number: BBI-8854600

SS CLAMP FOR FERNBACH 1800ML

UNSPSC: 41121710

Code: NEW

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 11730406

Quantity Price
1 £ 54.96 / Each
Estimated Shipment
07-04-2017
Add to basket


Description and Specification

Specification

Material Stainless Steel
Type Clamp
Capacity (Metric) 1.8L
Holds 1800mL Fernbach Flask
For Use With Certomat™ MOII RM SII IS, BS-1, BS-T, CT Plus