Thermo Scientific™ TurboFect Transfection Reagent

Get efficient in vitro delivery of plasmid DNA to cultured cells with low cytotoxicity.

Overview
Brand: Thermo Scientific™

Manufacturer Part Number: R0531

UNSPSC: 41116133

Code: 88

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 10304780

 N/A
N/A

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


  • Description and Specification
  • Alternative Products
Description and Specification

Specification

For Use With (Application) Transfection
Final Product Type TurboFect™ Transfection Reagent
Product Type TurboFect™ Transfection Reagent
Description TurboFect Transfection Reagent, 1mL, Liquid, Plasmid DNA Sample Type, Chemical Transfection Technique, Water Soluble, Used in Transfection, Stable Under Normal Conditions, Chemically Defined Classification
Formulation Liquid
Sample Type Plasmid DNA
Quantity 1mL

The polymer forms compact, stable, positively charged complexes with DNA. These complexes protect DNA from degradation and facilitate gene delivery in vivo. Reagent causes no detectable inflamatory response and is suitable for DNA administration via various routes, including intravenous, intraperitoneal, intratumoral injection, etc.
  • No detectable inflamatory response
  • Gene delivery via multiple routes of administration
  • Reproducible results
  • Functionally tested in mice and rats

Recommended for:

DNA transfection

siRNA-DNA co-transfection

Alternative Products